Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dipropylene Glycol Methyl Ether Acetate, 98%, Spectrum™ Chemical
SDP

Catalog No. 18610295 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
250 mL
1 L
4 L
20 L
Packaging:
Amber Glass Bottle
Steel Can
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-610-295 1 L Amber Glass Bottle
18-610-294 250 mL Amber Glass Bottle
18-610-296 4 L Amber Glass Bottle
18-610-297 20 L Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-610-295 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. D11111LT
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 88917-22-0
Molecular Formula C9H18O4
Molecular Weight (g/mol) 190.24
InChI Key USDXBNSCKZJDLC-UHFFFAOYNA-N
IUPAC Name 1-(3-methoxypropoxy)propyl acetate
SMILES CCC(OCCCOC)OC(C)=O

Specifications

Acidity 0.0005
CAS Max % 1
Packaging Amber Glass Bottle
Quantity 1 L
Percent Purity 0.98
Grade Reagent

CAUTION: For use by qualified personnel for research and development or other authorized use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.