Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DIRAS2 Antibody (OTI10H1), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP271967V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271967V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271967V Supplier Novus Biologicals Supplier No. NBP271967V

Mouse Monoclonal Antibody

DIRAS2 Monoclonal specifically detects DIRAS2 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen DIRAS2
Applications Western Blot
Classification Monoclonal
Clone OTI10H1
Conjugate DyLight 405
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias DIRAS family, GTP-binding RAS-like 2, Di-Ras2, Distinct subgroup of the Ras family member 2, DKFZp761C07121, GTP-binding protein Di-Ras2
Gene Symbols DIRAS2
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-196 of human DIRAS2 (NP_060064) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 54769.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.