Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DIRAS2 Antibody (OTI10H1), FITC, Novus Biologicals™
SDP

Catalog No. NB288510 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288510 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288510 Supplier Novus Biologicals Supplier No. NBP271967F

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

DIRAS2 Monoclonal antibody specifically detects DIRAS2 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen DIRAS2
Applications Western Blot
Classification Monoclonal
Clone OTI10H1
Conjugate FITC
Dilution Western Blot
Formulation PBS
Gene Alias DIRAS family, GTP-binding RAS-like 2, Di-Ras2, Distinct subgroup of the Ras family member 2, DKFZp761C07121, GTP-binding protein Di-Ras2
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-196 of human DIRAS2(NP_060064) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 54769
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.