Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DIRAS3 Antibody, DyLight 680, Novus Biologicals™
SDP

Catalog No. NB184931 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB184931 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB184931 Supplier Novus Biologicals Supplier No. NBP282085FR
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DIRAS3 Polyclonal antibody specifically detects DIRAS3 in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen DIRAS3
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias DIRAS family, GTP-binding RAS-like 3, Distinct subgroup of the Ras family member 3, NOEY2member I, RHOI, Rho-related GTP-binding protein RhoI
Host Species Rabbit
Immunogen DIRAS3 antibody was raised against an 18 amino acid peptide near the carboxy terminus of human DIRAS3. The immunogen is located within the last 50 amino acids of DIRAS3. Amino Acid Squence: TTGLQEPEKKSQMPNTTE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 9077
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.