Learn More
Invitrogen™ DIRAS3 Polyclonal Antibody

Description
Antibody detects endogenous levels of total DIRAS3.
Specifications
Specifications
| Antigen | DIRAS3 |
| Applications | Immunohistochemistry (PFA fixed), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide |
| Gene | DIRAS3 |
| Gene Accession No. | O95661 |
| Gene Alias | ARHI; DIRAS family GTP binding RAS like 3; DIRAS family GTPase 3; DIRAS family, GTP-binding RAS-like 3; DIRAS3; Distinct subgroup of the Ras family member 3; GTP-binding protein Di-Ras3; NOEY2; ras homolog gene family, member I; RGD1565168; RHOI; rho-related GTP-binding protein RhoI |
| Gene Symbols | DIRAS3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.