Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DISC1 Antibody, Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB11040775X Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11040775X 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11040775X Supplier Novus Biologicals Supplier No. NB11040775AF488
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DISC1 Polyclonal specifically detects DISC1 in Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen DISC1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Alexa Fluor 488
Dilution Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias C1orf136, disrupted in schizophrenia 1, disrupted in schizophrenia 1 protein, FLJ13381, FLJ21640, FLJ25311, FLJ41105, KIAA0457, SCZD9
Gene Symbols DISC1
Host Species Rabbit
Immunogen A synthetic peptide made to an internal region (within residues 700-800) of the mouse DISC1 protein. [Swiss-Prot# Q811T9]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Neurodegeneration, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 27185
Target Species Mouse, Rat
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.