Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BrandTech™ BRAND™ Discharge Tubes for Dispensette™ S Bottletop Dispensers

For use with Dispensette S Bottletop Dispensers.

$38.60 - $187.70

Specifications

Type Discharge Tube
Volume (Metric) 5 and 10 mL
For Use With (Equipment) Dispensette S Bottletop Dispensers
Tip Type Fine tip
View More Specs

Products 2
Catalog Number Mfr. No. Includes Price Quantity & Availability  
Catalog Number Mfr. No. Includes Price Quantity & Availability  
13-882-270
View Documents
BrandTech™ BRAND™
708012
Without recirculation valve
Each for $38.60
Add to cart
 
13-882-271
View Documents
BrandTech™ BRAND™
708112
Recirculation valve
Each for $187.70
Add to cart
 
Description

Description

  • Fine tip
  • Volume: 5 and 10mL
  • Available with or without recirculation valve
Specifications

Specifications

Discharge Tube
5 and 10 mL
Dispensette S Bottletop Dispensers
Fine tip
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit