Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Discovery™ BIO Wide Pore C18 HPLC Column, 3 μm Particle Size

Catalog No. 111003317 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
5 cm
10 cm
15 cm
Diameter (Metric):
2.1 mm
4.6 mm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
11-100-3317 10 cm 4.6 mm
11-100-3463 5 cm 2.1 mm
11-100-3316 10 cm 2.1 mm
11-100-3180 15 cm 2.1 mm
11-101-6173 15 cm 4.6 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 11-100-3317 Supplier MilliporeSigma™ Supelco™ Supplier No. 567204U
Add to Cart
Add to Cart

The Discovery™ BIO Wide Pore C18 column has the ability to analyze peptides without TFA in the mobile phase, thereby increasing the LC-MS signal.

Peptide maps generated by RP-HPLC provide valuable information about protein structure, stability, and purity. To be effective, the RP-HPLC column must be able to resolve a high percentage of the peptides in the sample. The more peptides, the better the information. Discovery™ BIO Wide Pore C18 gives unsurpassed RP-HPLC resolution of peptide maps from enzymatic digests. The improvements in silica and bonded-phase chemistry that have been incorporated into the Discovery™ BIO Wide Pore line improve resolution by increasing efficiency and reducing the peak tailing. An added benefit to this is the ability to analyze peptides without TFA in the mobile phase, thereby increasing the LC-MS signal.
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 4.6 mm
Length (Metric) 10 cm
Particle Size 3 μm
Pore Size 300 Å
Surface Area 100 m2/g
Stationary Phase C18 (octadecyl)
Product Line Discovery
Matrix C18 (octadecyl) Phase/silica particle platform
Endcapped Yes
Phase Reversed Phase
Technique Wide Pore HPLC
Show More Show Less

Discovery is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.