Learn More
MilliporeSigma™ Supelco™ Discovery™ C18 HPLC Column, 5 μm Particle Size
Shop All MilliporeSigma Supelco ProductsDescription
- Excellent reproducibility
- Exceptional peak shape for basic and acidic analytes
- Stable, low-bleed LC-MS separations
- Separation of peptides and small proteins
- Lower hydrophobicity than many comparable C18 columns, providing faster analysis
Specifications
Specifications
| Column Format | HPLC, LC/MS Column |
| Diameter (Metric) | 2.1 mm |
| Length (Metric) | 10 cm |
| Max. Pressure | 400 bar(5801 psi) |
| Particle Size | 5 μm |
| pH | 2 to 8 |
| Pore Size | 180 Å |
| Surface Area | 200 m2/g |
| Temperature | ≤70°C |
| USP Type | USP L1 |
| Show More |
Discovery is a registered trademark of Sigma-Aldrich Co. LLC
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.