Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dishevelled-1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP224648SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224648SS 0.025 mg
NBP224648 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224648SS Supplier Novus Biologicals Supplier No. NBP224648SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Dishevelled-1 Polyclonal specifically detects Dishevelled-1 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot.

Specifications

Antigen Dishevelled-1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 5-7 ug/ml
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. O14640
Gene Alias dishevelled 1 (homologous to Drosophila dsh), dishevelled, dsh homolog 1 (Drosophila), Dishevelled-1, DSH homolog 1, DVL, DVL1L1, MGC54245, segment polarity protein dishevelled homolog DVL-1
Gene Symbols DVL1
Host Species Rabbit
Immunogen A portion of amino acids 70-120 of human DVL-1 was used as the immunogen.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 1855
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, rhesus monkey, pig, opossum, chimpanzee, dog and chicken, and 94% homologous in rat and mouse.
Target Species Human, Mouse, Rat, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.