Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dishevelled-1 Antibody, Novus Biologicals™
SDP

Catalog No. NB036584 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB036584 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB036584 Supplier Novus Biologicals Supplier No. NBP288993
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Dishevelled-1 Polyclonal specifically detects Dishevelled-1 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen Dishevelled-1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, pH 7.4
Gene Alias dishevelled 1 (homologous to Drosophila dsh), dishevelled, dsh homolog 1 (Drosophila), Dishevelled-1, DSH homolog 1, DVL, DVL1L1, MGC54245, segment polarity protein dishevelled homolog DVL-1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to aa 557-571 (CS557RRAPGREKERRAAG571) of human Dishevelled-1 protein.
Purification Method Protein A purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 1855
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.