Learn More
Dishevelled-2 Antibody (OTI2D11), Alexa Fluor™ 405, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Dishevelled-2 |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Clone | OTI2D11 |
| Conjugate | Alexa Fluor 405 |
| Dilution | Immunohistochemistry |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | dishevelled 2 (homologous to Drosophila dsh), dishevelled, dsh homolog 2 (Drosophila), Dishevelled-2, DSH homolog 2, segment polarity protein dishevelled homolog DVL-2 |
| Gene Symbols | DVL2 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.