Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DISP2 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB341875 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB341875 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB341875 Supplier Novus Biologicals Supplier No. NBP281918MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DISP2 Polyclonal antibody specifically detects DISP2 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen DISP2
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias Dispatched B, Dispatched Homolog 2 (Drosophila), DISPB, HsT16908, KIAA1742, Protein Dispatched Homolog 2
Host Species Rabbit
Immunogen DISP2 antibody was raised against an 18 amino acid synthetic peptide near the amino terminus of human DISP2. The immunogen is located within amino acids 110 - 160 of DISP2. Amino Acid Squence: SPSPAPSQRDGTWKPPA
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 85455
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.