Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Distance Learning: Cellular Respiration- What Sugar Does Yeast Like Best
GREENER_CHOICESCI_ED

Catalog No. S08201 Shop All Aldon Products
Change view
Click to view available options
Grade:
7 to 15
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S08201 7 to 15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S08201 Supplier Innovating Science™ Supplier No. IS1510
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Allows students to understand yeast may use different options for energy production with varying degrees of effectiveness

  • Objectives:
    • Expose living yeast cells to glucose, sucrose, and lactose
    • Observe yeast using sugar as food source
    • Allows students to understand yeast can use different options for energy production
    • Use pH indicator to indirectly determine the effectiveness of each sugar
  • Includes:
    • 10 mL 0.5% litmus
    • 1 g dehydrated yeast
    • 5 mL 0.1M sodium hydroxide
    • 1 glucose QuickSolution (to make 30 mL)
    • 1 sucrose QuickSolution (to make 30 mL)
    • 1 lactose QuickSolution (to make 30 mL)
    • 6 plastic cups
    • 2 plastic scoops
    • 2 pipettes
  • Notes:
    • Contains and an instruction manual and enough materials for two groups

Specifications

Grade 7 to 15
Age 12 to 20
Class Size 2 Lab Groups
Product Type Cellular Respiration Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.