Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Distemper Virus Antibody (8-1), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB1102603IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1102603IR 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1102603IR Supplier Novus Biologicals Supplier No. NB1102603IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Distemper Virus Monoclonal specifically detects Distemper Virus in Virus samples. It is validated for ELISA.

Specifications

Antigen Distemper Virus
Applications ELISA
Classification Monoclonal
Clone 43313
Conjugate DyLight 755
Dilution ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Canine distemper virus (CDV).
Purification Method Protein A purified
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Specific for surface envelope antigen of CDV
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.