Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Dithizone
SCI_ED

Catalog No. s25685 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
2.5 g
Packaging:
Solid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25685 2.5 g Solid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25685
Only null left
Add to Cart
Add to Cart

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 60-10-6
Molecular Formula C13H12N4S
Molecular Weight (g/mol) 256.33
MDL Number MFCD00003025
InChI Key UOFGSWVZMUXXIY-UHFFFAOYSA-N
Synonym dithizone, dithizon, ditizon, carbazone, diphenylthio, usaf ek-3092, diazenecarbothioic acid, phenyl-, 2-phenylhydrazide, 1,5-diphenyl-3-mercaptoformazan, 3-formazanthiol, 1,5-diphenyl, unii-njz2cj4d6p, 1,5-diphenyl-3-thiocarbazone
PubChem CID 657262
IUPAC Name 3-(phenylamino)-1-(phenylimino)thiourea
SMILES S=C(NNC1=CC=CC=C1)N=NC1=CC=CC=C1

Specifications

Packaging Solid
Linear Formula C13H12N4S
Quantity 2.5 g
Formula Weight 256.32
Chemical Name or Material Dithizone
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.