Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DKFZP434B168 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB291974 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291974 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291974 Supplier Novus Biologicals Supplier No. NB100465PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DKFZP434B168 Polyclonal specifically detects DKFZP434B168 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen DKFZP434B168
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias C1orf73, chromosome 1 open reading frame 73, DKFZp434B168, Int7, integrator complex subunit 7
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residues 850-900 of human DKFZP434B168 using the TrEMBL entry Q9NVH2 (GeneID 25896).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 25896
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.