Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DKFZp566F084 Antibody, Novus Biologicals™
SDP

Catalog No. NBP15520020 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15520020 20 μL
NBP155200 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15520020 Supplier Novus Biologicals Supplier No. NBP15520020UL

Rabbit Polyclonal Antibody

DKFZp566F084 Polyclonal specifically detects DKFZp566F084 in Human samples. It is validated for Western Blot.

Specifications

Antigen DKFZp566F084
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q9BVM2
Gene Alias deleted in a mouse model of primary ciliary dyskinesia, deleted in primary ciliary dyskinesia homolog (mouse), DKFZP566F084, protein DPCD, RP11-529I10.4
Gene Symbols DPCD
Host Species Rabbit
Immunogen Synthetic peptides corresponding to RP11-529I10.4(deleted in a mouse model of primary ciliary dyskinesia) The peptide sequence was selected from the N terminal of RP11-529I10.4. Peptide sequence MAEEYDEKTSELLVRKWRVKSALGAMGQWQLEVGDPAPLGAGNL
Molecular Weight of Antigen 23 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25911
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.