Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DKFZp566F084 Antibody (OTI4B9), Biotin, Novus Biologicals™
SDP

Catalog No. NBP271779B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271779B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271779B Supplier Novus Biologicals Supplier No. NBP271779B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DKFZp566F084 Monoclonal specifically detects DKFZp566F084 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen DKFZp566F084
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI4B9
Conjugate Biotin
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.05% Sodium Azide
Gene Alias deleted in a mouse model of primary ciliary dyskinesia, deleted in primary ciliary dyskinesia homolog (mouse), DKFZP566F084, protein DPCD, RP11-529I10.4
Gene Symbols DPCD
Host Species Mouse
Immunogen Full length recombinant protein of human DKFZp566F084 (NP_056263) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 25911.0
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.