Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dkk-1 Antibody (004), Biotin, Novus Biologicals™
SDP

Catalog No. NB229416 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB229416 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB229416 Supplier Novus Biologicals Supplier No. NBP289309B
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Dkk-1 Monoclonal antibody specifically detects Dkk-1 in Human samples. It is validated for ELISA

Specifications

Antigen Dkk-1
Applications ELISA
Classification Monoclonal
Clone 004
Conjugate Biotin
Dilution ELISA
Formulation PBS
Gene Alias dickkopf (Xenopus laevis) homolog 1, dickkopf homolog 1 (Xenopus laevis), dickkopf related protein-1, Dickkopf-1, dickkopf-related protein 1, DKK-1, hDkk-1, SKdickkopf-1 like
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Dkk-1 (Met 2-His 266; NP_036374.1).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity, Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 22943
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.