Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dkk-1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP224701SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224701SS 0.025 mg
NBP224701 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224701SS Supplier Novus Biologicals Supplier No. NBP224701SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Dkk-1 Polyclonal specifically detects Dkk-1 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen Dkk-1
Applications Western Blot
Classification Polyclonal
Concentration 1.0 mg/ml
Conjugate Unconjugated
Dilution Western Blot 0.5-2.0 ug/ml
Formulation PBS with 0.05% Sodium Azide
Gene Alias dickkopf (Xenopus laevis) homolog 1, dickkopf homolog 1 (Xenopus laevis), dickkopf related protein-1, Dickkopf-1, dickkopf-related protein 1, DKK-1, hDkk-1, SKdickkopf-1 like
Gene Symbols DKK1
Host Species Rabbit
Immunogen A portion of amino acids 150-200 of human DKK1 was used as the immunogen.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity, Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 22943
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, 97% homologous in mouse, dog and horse, 95% homologous in rat and 94% homologous in cow.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.