Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dkk-2 Antibody (994930) [DyLight 550], Novus Biologicals™
SDP

Catalog No. NB376121 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB376121 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB376121 Supplier Novus Biologicals Supplier No. FAB6628L
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Dkk-2 Monoclonal antibody specifically detects Dkk-2 in Human samples. It is validated for Immunohistochemistry, Flow Cytometry (Intracellular Staining), Immunocytochemistry

Specifications

Antigen Dkk-2
Applications Immunohistochemistry, Flow Cytometry (Intracellular Staining), Immunocytochemistry
Classification Monoclonal
Clone 994930
Conjugate DyLight 550
Dilution Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry
Formulation 50mM Sodium Borate
Gene Alias dickkopf (Xenopus laevis) homolog 2, dickkopf 2, dickkopf homolog 2 (Xenopus laevis), dickkopf related protein-2, dickkopf-2, dickkopf-related protein 2, DKK-2, hDkk-2
Host Species Mouse
Immunogen Chinese Hamster Ovary cell line, CHO-derived human Dkk-2, Met1-Ile259, Accession # NP_055236
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 27123
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.