Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dkk-3 Antibody (103B5), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB162976 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB162976 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB162976 Supplier Novus Biologicals Supplier No. NBP260242V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Dkk-3 Monoclonal antibody specifically detects Dkk-3 in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen Dkk-3
Applications Western Blot, ELISA
Classification Monoclonal
Clone 103B5
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias dickkopf (Xenopus laevis) homolog 3, dickkopf 3, dickkopf homolog 3 (Xenopus laevis), Dickkopf-3, dickkopf-related protein 3, Dkk-3, hDkk-3, regulated in glioma, REICdkk-3, RIG, RIG-like 5-6, RIG-like 7-1
Host Species Mouse
Immunogen Recombinant human DKK-3 (22-350aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 27122
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.