Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dkk-3 Mouse anti-Human, Mouse, Clone: 103B5, Novus Biologicals™
SDP

Catalog No. NB60242100U Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB60242100U 100 μL
NB6024250UL 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB60242100U Supplier Novus Biologicals Supplier No. NBP260242100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Dkk-3 Monoclonal specifically detects Dkk-3 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Dkk-3
Applications Western Blot, ELISA
Classification Monoclonal
Clone 103B5
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, ELISA 1:100 - 1:2000
Formulation PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
Gene Alias dickkopf (Xenopus laevis) homolog 3, dickkopf 3, dickkopf homolog 3 (Xenopus laevis), Dickkopf-3, dickkopf-related protein 3, Dkk-3, hDkk-3, regulated in glioma, REICdkk-3, RIG, RIG-like 5-6, RIG-like 7-1
Gene Symbols DKK3
Host Species Mouse
Immunogen Recombinant human DKK3 (22-350aa) purified from E. coli
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 27122
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.