Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dkk-3 Mouse anti-Human, Mouse, Clone: 103B5, Novus Biologicals™

Mouse Monoclonal Antibody
$340.50 - $499.50
Specifications
Antigen | Dkk-3 |
---|---|
Clone | 103B5 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:1000, ELISA 1:100 - 1:2000 |
Applications | Western Blot, ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB6024250UL
![]() |
Novus Biologicals
NBP26024250UL |
50 μL |
Each for $340.50
|
|
|||||
NB60242100U
![]() |
Novus Biologicals
NBP260242100UL |
100 μL |
Each for $499.50
|
|
|||||
Description
Dkk-3 Monoclonal specifically detects Dkk-3 in Human samples. It is validated for Western Blot, ELISA.Specifications
Dkk-3 | |
1 mg/mL | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
Human, Mouse | |
dickkopf (Xenopus laevis) homolog 3, dickkopf 3, dickkopf homolog 3 (Xenopus laevis), Dickkopf-3, dickkopf-related protein 3, Dkk-3, hDkk-3, regulated in glioma, REICdkk-3, RIG, RIG-like 5-6, RIG-like 7-1 | |
DKK3 | |
IgG1 κ | |
Protein A purified |
103B5 | |
Western Blot 1:1000, ELISA 1:100 - 1:2000 | |
Monoclonal | |
Purified | |
Signal Transduction | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
27122 | |
Recombinant human DKK3 (22-350aa) purified from E. coli | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title