Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dkk-4 Antibody, Novus Biologicals™
SDP

Catalog No. NB1001000 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001000 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001000 Supplier Novus Biologicals Supplier No. NB1001000
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Dkk-4 Polyclonal specifically detects Dkk-4 in Human samples. It is validated for Peptide ELISA, Western Blot.

Specifications

Antigen Dkk-4
Applications Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. Q9UBT3, NP_055235.1
Gene Alias dickkopf (Xenopus laevis) homolog 4, dickkopf homolog 4 (Xenopus laevis), dickkopf-4, dickkopf-related protein 4, DKK-4, hDkk-4, MGC129562, MGC129563
Gene Symbols DKK4
Host Species Goat
Immunogen Peptide with sequence RQHARLRVCQKIEKL corresponding to C-Terminus according to NP_055235.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 27121
Test Specificity Human. Predicted cross-reactivity based on sequence identity: Mouse, Rat, Dog, Pig.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.