Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DL-Cystine
Click to view available options
Quantity:
5 g
25 g
100 g
Description
DL-cystine is used in the preparation of sulfur-containing dimeric and monomeric surfactants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
DL-cystine is used in the preparation of sulfur-containing dimeric and monomeric surfactants.
Solubility
Soluble in dilute acid and alkali solution, highly soluble in water, not soluble in alcohol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
DL-cystine is used in the preparation of sulfur-containing dimeric and monomeric surfactants.
Solubility
Soluble in dilute acid and alkali solution, highly soluble in water, not soluble in alcohol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications
Specifications
| Melting Point | 227°C (decomposition) |
| Color | White |
| Beilstein | 1728095 |
| Merck Index | 14,2782 |
| Solubility Information | Soluble in dilute acid and alkali solution,highly soluble in water,not soluble in alcohol. |
| Quantity | 25 g |
| Formula Weight | 240.3 |
| Physical Form | Powder |
| Chemical Name or Material | DL-Cystine |
Safety and Handling
| Hazard Category | H302-H315-H319-H335 |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| EINECSNumber | 213-094-5 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction