Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Ethyl 2-bromovalerate 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
615-83-8
C7H13BrO2
209.08
MFCD00000159
ORSIRXYHFPHWTN-UHFFFAOYSA-N
ethyl 2-bromovalerate,ethyl2-bromovalerate,ethyl .alpha.-bromovalerate,pentanoic acid, 2-bromo-, ethyl ester,2-bromovaleric acid ethyl ester,2-bromopentanoic acid ethyl ester,2-bromo-pentanoic acid ethyl ester,pentanoic acid, bromo-, ethyl ester,ethyl bromovalerate,ethyl-2bromovalerate
79071
ethyl 2-bromopentanoate
CCCC(C(=O)OCC)Br
615-83-8
C7H13BrO2
209.08
MFCD00000159
ORSIRXYHFPHWTN-UHFFFAOYSA-N
ethyl 2-bromovalerate,ethyl2-bromovalerate,ethyl .alpha.-bromovalerate,pentanoic acid, 2-bromo-, ethyl ester,2-bromovaleric acid ethyl ester,2-bromopentanoic acid ethyl ester,2-bromo-pentanoic acid ethyl ester,pentanoic acid, bromo-, ethyl ester,ethyl bromovalerate,ethyl-2bromovalerate
79071
ethyl 2-bromopentanoate
CCCC(C(=O)OCC)Br
Refractive Index 1.4476 to 1.4496
Color Colorless
Boiling Point 190.0°C to 192.0°C
Packaging Glass bottle
Flash Point 77°C
Infrared Spectrum Authentic
Density 1.2200g/mL
Specific Gravity 1.22
Quantity 25g
Assay Percent Range 99%
Linear Formula CH3CH2CH2CH(Br)CO2C2H5
Beilstein 02, 302
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Formula Weight 209.08
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material DL-Ethyl 2-bromovalerate, 99%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

LACHRYMATOR

EINECSNumber : 210-450-1