Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Glyceraldehyde Diethyl Acetal 95.0+%, TCI America™
SDP

Catalog No. G021625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G021625G 25 g
G02165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G021625G Supplier TCI America Supplier No. G021625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10487-05-5
Molecular Formula C7H16O4
Molecular Weight (g/mol) 164.201
MDL Number MFCD00059599
InChI Key BPNZBKXZVJVEJD-UHFFFAOYSA-N
PubChem CID 542749
IUPAC Name 3,3-diethoxypropane-1,2-diol
SMILES CCOC(C(CO)O)OCC

Specifications

Color Colorless
Boiling Point 136°C
Quantity 25 g
Formula Weight 164.20
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material DL-Glyceraldehyde Diethyl Acetal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.