Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Methionine sulfone, 98%

Catalog No. p-7027993
Click to view available options
Quantity:
1 g
5 g

CAS: 820-10-0 | C5H11NO4S | 181.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 820-10-0
Molecular Formula C5H11NO4S
Molecular Weight (g/mol) 181.21
MDL Number MFCD00025079
InChI Key UCUNFLYVYCGDHP-UHFFFAOYNA-N
Synonym dl-methionine sulfone, methionine sulfone, 2-amino-4-methylsulfonyl butanoic acid, butanoic acid, 2-amino-4-methylsulfonyl, d,l-methionine sulfone, methioninesulfone, dl-2-amino-4-methylsulfonyl butanoic acid, 2-amino-4-methylsulfonyl butyric acid, methionine, s,s-dioxide, butyric acid, 2-amino-4-methylsulfonyl-, dl
PubChem CID 69961
IUPAC Name 2-amino-4-methylsulfonylbutanoic acid
SMILES CS(=O)(=O)CCC(N)C(O)=O

Specifications

Quantity 1 g
Formula Weight 181.21
Percent Purity 98%
Chemical Name or Material DL-Methionine sulfone
EINECSNumber 212-466-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.