Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-alpha-Tocopherol, 97+%

Catalog No. AAA170390B
Click to view available options
Quantity:
50 g
250 g
1000 g

Catalog No. AAA170390B

Supplier: Thermo Scientific Chemicals A170390B

Only null left
Add to Cart

CAS: 10191-41-0 | C29H50O2 | 430.72 g/mol

DL-α-Tocopherol is used as an antioxidant that protects cell membrane lipids from oxidative damage.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-α-Tocopherol is used as an antioxidant that protects cell membrane lipids from oxidative damage.

Solubility
Miscible with chloroform, vegetable oils,  ether, acetone and alcohol. Immiscible with water.

Notes
Store in a cool place. Incompatible withstrong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10191-41-0
Molecular Formula C29H50O2
Molecular Weight (g/mol) 430.72
MDL Number MFCD00072051
InChI Key GVJHHUAWPYXKBD-IEOSBIPESA-N
Synonym vitamin e, alpha-tocopherol, d-alpha-tocopherol, 5,7,8-trimethyltocol, +-alpha-tocopherol, r,r,r-alpha-tocopherol, phytogermine, eprolin, 2r,4'r,8'r-alpha-tocopherol, dl-a-tocopherol
PubChem CID 14985
ChEBI CHEBI:18145
IUPAC Name (2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-ol
SMILES CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=C(C)C(O)=C(C)C(C)=C2O1

Specifications

Melting Point 2°C to 4°C
Density 0.95
Boiling Point 200°C to 220°C (0.1 mmHg)
Flash Point 240°C (464°F)
Assay Percent Range >97%
Refractive Index 1.505
Quantity 1000 g
Beilstein 94012
Sensitivity Air and light sensitive
Merck Index 14,10021
Solubility Information Miscible with chloroform,vegetable oils, ether,acetone and alcohol. Immiscible with water.
Formula Weight 430.72
Percent Purity ≥97%
Chemical Name or Material DL-alpha-Tocopherol
Show More Show Less
Hazard Category H317
Precautionary Statement P261-P272-P280g-P302+P352-P333+P313-P363-P501c
EINECSNumber 233-466-0
RTECSNumber GA8746000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.