Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DLC1 Antibody - Isoform 1 and 3, Novus Biologicals™
SDP

Catalog No. NB300917 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300917 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300917 Supplier Novus Biologicals Supplier No. NB300917
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 2 publications

DLC1 Polyclonal specifically detects DLC1 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.

Specifications

Antigen DLC1 (Isoform 1 and 3)
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry 4 ug/mL, Immunohistochemistry-Paraffin 4 ug/mL, Immunofluorescence 10 ug/mL
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_079043
Gene Alias rho GTPase-activating protein 7, ARHGAP7, deleted in liver cancer 1, HP, p122-RhoGAP, STARD12
Gene Symbols DLC1
Host Species Goat
Immunogen Peptide with sequence SVAIRKRSWEEHC corresponding to N-Terminus according to NP_872584.2, NP_079043.3.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10395
Test Specificity This antibody is expected to recognize isoforms 1 and 3 ( NP_872584.2 and NP_079043.3 resp) but NOT isoform 2 (NP_006085.2).
Target Species Human, Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.