Learn More
Description
Specifications
Specifications
| Antigen | DLC1/DLEC1 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 532 |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate |
| Gene Alias | DLC1, DLEC1, DLEC1 deleted in lung and esophageal cancer 1, F56 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human DLC1/DLEC1 fragment. |
| Purification Method | Antigen and protein A Affinity-purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
