Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DLG1 Antibody (RPI 197.4), Novus Biologicals™
SDP

Catalog No. NB036585 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
200 ug
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB036585 200 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB036585 Supplier Novus Biologicals Supplier No. NBP288994
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DLG1 Monoclonal specifically detects DLG1 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Electron Microscopy.

Specifications

Antigen DLG1
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunomicroscopy
Classification Monoclonal
Clone RPI 197.4
Conjugate Unconjugated
Formulation PBS, pH 7.2, containing 50% glycerol
Gene Alias discs, large homolog 1 (Drosophila), disks large homolog 1, DKFZp761P0818, DKFZp781B0426, hDlg, large (Drosophila) homolog 1, presynaptic protein SAP97
Host Species Mouse
Immunogen Recombinant rat DLG1(N-terminal fragment).
Purification Method Protein G purified
Quantity 200 ug
Regulatory Status RUO
Research Discipline Neuroscience, Vision
Primary or Secondary Primary
Gene ID (Entrez) 1739
Target Species Human, Mouse, Rat, Canine
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.