Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DLGAP3 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB163870 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB163870 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB163870 Supplier Novus Biologicals Supplier No. NBP281713F
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DLGAP3 Polyclonal antibody specifically detects DLGAP3 in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen DLGAP3
Applications ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias DAP-3, discs, large (Drosophila) homolog-associated protein 3, disks large-associated protein 3, SAP90/PSD 95 associated protein 3, SAP90/PSD-95-associated protein 3
Host Species Rabbit
Immunogen DLGAP3 antibody was raised against a 18 amino acid synthetic peptide from near the carboxy terminus of human DLGAP3. The immunogen is located within the last 50 amino acids of DLGAP3. Amino Acid Squence: KERSLDSVDRQRQEARKR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 58512
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.