Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DLL1 Antibody (019), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB075713 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB075713 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB075713 Supplier Novus Biologicals Supplier No. NBP290620AF647
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

DLL1 Monoclonal specifically detects DLL1 in Mouse samples. It is validated for ELISA.

Specifications

Antigen DLL1
Applications ELISA
Classification Monoclonal
Clone 19
Conjugate Alexa Fluor 647
Formulation 50 mM Sodium Borate
Gene Alias Delta, delta (Drosophila)-like 1, Delta1, delta-like 1 (Drosophila), delta-like protein 1, DL1, Drosophila Delta homolog 1, H-Delta-1
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse DLL1 (NP_031891.2; Met 1-Gln 516).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience, Stem Cell Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 28514
Test Specificity Has cross-reactivity in ELISA with: Human DLL1
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.