Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DLL1 Antibody (MM0247- 2X38), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP212266FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212266FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP212266FR Supplier Novus Biologicals Supplier No. NBP212266FR

Mouse Monoclonal Antibody

DLL1 Monoclonal specifically detects DLL1 in Human samples. It is validated for Western Blot.

Specifications

Antigen DLL1
Applications Western Blot
Classification Monoclonal
Clone MM0247- 2X38
Conjugate DyLight 680
Dilution Western Blot
Gene Accession No. O00548
Gene Alias Delta, delta (Drosophila)-like 1, Delta1, delta-like 1 (Drosophila), delta-like protein 1, DL1, Drosophila Delta homolog 1, H-Delta-1
Gene Symbols DLL1
Host Species Mouse
Immunogen Human recombinant DLL1
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience, Stem Cell Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 28514
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.