Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DLL3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP224669 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224669 0.1 mg
NBP224669SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224669 Supplier Novus Biologicals Supplier No. NBP224669
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

DLL3 Polyclonal specifically detects DLL3 in Human, Mouse, Primate samples. It is validated for IF/IHC, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen DLL3
Applications Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 3 - 5 ug/mL, Immunohistochemistry reported in scientific literature (PMID 31109352), Immunohistochemistry-Paraffin 5 ug/mL
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. Q9NYJ7
Gene Alias delta (Drosophila)-like 3, Delta3, delta-like 3 (Drosophila), delta-like protein 3, Drosophila Delta homolog 3, SCDO1delta3
Gene Symbols DLL3
Host Species Rabbit
Immunogen A portion of amino acid 100 to 150 of human DLL3 was used as immunogen for the antibody.
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10683
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human and monkey, 83% homologous in cat, 81% homologous in bovine and 72% homologous in mouse and rat. IMG-6242A detects the expected approximately 23 kD band in mouse and rat skeletal muscle tissue lysate, but additional larger molecular weight bands may be seen.
Target Species Human, Mouse, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.