Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DLL3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$206.50 - $512.00
Specifications
| Antigen | DLL3 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 3 - 5 ug/mL, Immunohistochemistry reported in scientific literature (PMID 31109352), Immunohistochemistry-Paraffin 5 ug/mL |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP224669SS
![]() |
Novus Biologicals
NBP224669SS |
0.025 mg |
Each for $206.50
|
|
|||||
NBP224669
![]() |
Novus Biologicals
NBP224669 |
0.1 mg |
Each for $512.00
|
|
|||||
Description
DLL3 Polyclonal specifically detects DLL3 in Human, Mouse, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| DLL3 | |
| Western Blot 3 - 5 ug/mL, Immunohistochemistry reported in scientific literature (PMID 31109352), Immunohistochemistry-Paraffin 5 ug/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 0.05% Sodium Azide | |
| delta (Drosophila)-like 3, Delta3, delta-like 3 (Drosophila), delta-like protein 3, Drosophila Delta homolog 3, SCDO1delta3 | |
| DLL3 | |
| IgG | |
| Protein A purified | |
| The amino acid sequence used as immunogen is 100% homologous in human and monkey, 83% homologous in cat, 81% homologous in bovine and 72% homologous in mouse and rat. IMG-6242A detects the expected approximately 23 kD band in mouse and rat skeletal muscle tissue lysate, but additional larger molecular weight bands may be seen. |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Primate | |
| Q9NYJ7 | |
| 10683 | |
| A portion of amino acid 100 to 150 of human DLL3 was used as immunogen for the antibody. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title