Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DLL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113841
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA5113841 100 μL
1 options

Catalog No. PIPA5113841

Supplier: Invitrogen™ PA5113841

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LVCACAPGYM GARCEFPVHP DGASALPAAP PGLRPGDPQR YLLPPALGLL For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 86%; Human: 100%; Mouse: 79%; Pig: 93%.

Delta-like protein 3 (DLL3) is a ligand for the Notch signaling pathway. It inhibits primary neurogenesis. DLL3 plays a role in the formation of somite boundaries during segmentation of the paraxial mesoderm. Defects in DLL3 are the cause of Spondylocostal dysostosis autosomal recessive type 1 (SCDO1). Mutations in DLL3 gene cause truncal shortening relative to their limbs, which leads to abdominal protrusion, abnormal spinal curvature and sometimes a plagiocephaly-torticollis sequence. It may be required to divert neurons along a specific differentiation pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DLL3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Dll3
Gene Accession No. Q9NYJ7
Gene Alias delta like canonical Notch ligand 3; Delta3; delta-like 3; delta-like 3 (Drosophila); delta-like protein 3; Dll3; Drosophila Delta homolog 3; M-Delta-3; pu; pudgy; SCDO1
Gene Symbols Dll3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal region of Human DLL3 (aa 451-500).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10683
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.