Learn More
Description
Specifications
Specifications
| Antigen | DLL4 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 3 |
| Conjugate | DyLight 550 |
| Dilution | ELISA, Sandwich ELISA Detection |
| Formulation | 50mM Sodium Borate |
| Gene Alias | delta 4, Delta 4 precursor, delta ligand 4, delta4, delta-like 4 (Drosophila), delta-like 4 homolog, delta-like 4 homolog (Drosophila), delta-like 4 protein, delta-like protein 4, Drosophila Delta homolog 4, hdelta2, MGC126344, notch ligand delta-2, notch ligand DLL4 |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human DLL4 extracellular domain (NP_061947.1; Met 1-Pro 524). |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
