Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DLL4 Antibody (447506), PE/Cy7, Novus Biologicals™
SDP

Catalog No. NB314477 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB314477 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB314477 Supplier Novus Biologicals Supplier No. FAB1506PECY7
Add to Cart
Add to Cart

Rat Monoclonal Antibody

DLL4 Monoclonal antibody specifically detects DLL4 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen DLL4
Applications Flow Cytometry
Classification Monoclonal
Clone 447506
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias delta 4, Delta 4 precursor, delta ligand 4, delta4, delta-like 4 (Drosophila), delta-like 4 homolog, delta-like 4 homolog (Drosophila), delta-like 4 protein, delta-like protein 4, Drosophila Delta homolog 4, hdelta2, MGC126344, notch ligand delta-2, notch ligand DLL4
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant human DLL4, Ser27-Pro524, Accession # Q9NR61
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 54567
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.