Learn More
Description
Specifications
Specifications
| Antigen | DIO3 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.5 ug/ml, Flow Cytometry reported by customer review using biotinylated antibody, Immunohistochemistry reported in scientific literature (PMID 24174657), Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 24885572), Immunohistochemistry-Paraffin reported in scientific literature (PMID 25004090), Immunohistochemistry-Frozen reported in scientific literature (PMID 24885572) |
| Formulation | PBS and 30% Glycerol with 0.1% Sodium Azide |
| Gene Alias | 5DIII, D3, deiodinase, iodothyronine, type III, DIOIII, EC 1.97.1, EC 1.97.1.11, ITDI3, placental type, thyroxine deiodinase type III (selenoprotein), Type 3 DI, type 3 iodothyronine selenodeiodinase, type III iodothyronine deiodinase, type-III 5' deiodinase, Type-III 5'-deiodinase |
| Gene Symbols | DIO3 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
