Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DIO3 Antibody - BSA Free, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 13 publications

$206.50 - $512.00

Specifications

Antigen DIO3
Concentration 1 mg/mL
Dilution Western Blot 0.5 ug/ml, Flow Cytometry reported by customer review using biotinylated antibody, Immunohistochemistry reported in scientific literature (PMID 24174657), Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 24885572), Immunohistochemistry-Paraffin reported in scientific literature (PMID 25004090), Immunohistochemistry-Frozen reported in scientific literature (PMID 24885572)
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP105767SS
SDP
View Documents
Novus Biologicals
NBP105767SS
0.025 mL
Each for $206.50
Only null left
Add to Cart
 
NBP105767
SDP
View Documents
Novus Biologicals
NBP105767
0.1 mL
Each for $512.00
Only null left
Add to Cart
 
Description

Description

DIO3 Polyclonal specifically detects DIO3 in Human, Mouse, Rat, Hamster, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

DIO3
Western Blot 0.5 ug/ml, Flow Cytometry reported by customer review using biotinylated antibody, Immunohistochemistry reported in scientific literature (PMID 24174657), Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 24885572), Immunohistochemistry-Paraffin reported in scientific literature (PMID 25004090), Immunohistochemistry-Frozen reported in scientific literature (PMID 24885572)
Polyclonal
Rabbit
Cancer, Growth and Development, Lipid and Metabolism, Neuroscience, Signal Transduction
PBS and 30% Glycerol with 0.1% Sodium Azide
1735
Synthetic peptide made to an internal portion of rat DIO3 (within residues 250-300). [Swiss-Prot# P49897]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
31 kDa
1 mg/mL
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human, Mouse, Rat, Hamster, Rabbit
5DIII, D3, deiodinase, iodothyronine, type III, DIOIII, EC 1.97.1, EC 1.97.1.11, ITDI3, placental type, thyroxine deiodinase type III (selenoprotein), Type 3 DI, type 3 iodothyronine selenodeiodinase, type III iodothyronine deiodinase, type-III 5' deiodinase, Type-III 5'-deiodinase
DIO3
IgG
Affinity Purified
Human, rat, hamster and mouse. Rabbit reactivity via customer review.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.