Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DLST Antibody, Novus Biologicals™
SDP

Catalog No. NBP232204 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP232204 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP232204 Supplier Novus Biologicals Supplier No. NBP232204
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DLST Polyclonal specifically detects DLST in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen DLST
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000 - 1:10000, Immunohistochemistry 1:500 - 1:2000, Immunoprecipitation 2 - 10 ug/mg lysate, Immunohistochemistry-Paraffin 1:500 - 1:2000
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_001924.2
Gene Alias 2-oxoglutarate dehydrogenase complex component E2, dihydrolipoamide S-succinyltransferase (E2 component of 2-oxo-glutaratecomplex), Dihydrolipoamide succinyltransferase component of 2-oxoglutarate dehydrogenasecomplex, dihydrolipoyllysine-residue succinyltransferase component of 2-oxoglutaratedehydrogenase complex, mitochondrial, DLTS, E2K, EC 2.3.1, EC 2.3.1.61, OGDC-E2
Gene Symbols DLST
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 50 to 100 of human Dihydrolipoamide S-succinyltransferase using the numbering given in entry NP_001924.2 (GeneID 1743).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1743
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.