Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dlx6 Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB044733 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB044733 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB044733 Supplier Novus Biologicals Supplier No. NBP297544AF750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Dlx6 Polyclonal specifically detects Dlx6 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Dlx6
Applications Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias distal-less homeo box 6, distal-less homeobox 6, homeobox protein DLX-6, MGC125282, MGC125283, MGC125284, MGC125285
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human Dlx6 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 1750
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.