Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DMBT1/GP340 Antibody (1G4) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP22357601 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP22357601 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP22357601 Supplier Novus Biologicals Supplier No. NBP2235760.1ML
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 2 publications

DMBT1/GP340 Monoclonal specifically detects DMBT1/GP340 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen DMBT1/GP340
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone 1G4
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500
Formulation 10mM Phosphate (pH 7.4) and 0.5M NaCl with 15mM Sodium Azide
Gene Alias deleted in malignant brain tumors 1, deleted in malignant brain tumors 1 protein, FLJ61058, Glycoprotein 340, gp-340, GP340muclin, Hensin, MGC164738, SAG, Salivary agglutinin, Surfactant pulmonary-associated D-binding protein
Gene Symbols DMBT1
Host Species Mouse
Immunogen Native gp-340 purified from bronchoalveolar lavage fluid from patients with pulmonary alveolar proteinosis, adsorbed onto aluminum hydroxide gel
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Breast Cancer
Primary or Secondary Primary
Gene ID (Entrez) 1755
Test Specificity Clone 1G4 Reacts with gp-340 and salivary agglutinin
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.