Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DMBT1/GP340 Antibody (548031), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB377300 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB377300 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB377300 Supplier Novus Biologicals Supplier No. FAB5915J
Add to Cart
Add to Cart

Rat Monoclonal Antibody

DMBT1/GP340 Monoclonal antibody specifically detects DMBT1/GP340 in Mouse samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen DMBT1/GP340
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 548031
Conjugate Janelia Fluor 646
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias deleted in malignant brain tumors 1, deleted in malignant brain tumors 1 protein, FLJ61058, Glycoprotein 340, gp-340, GP340muclin, Hensin, MGC164738, SAG, Salivary agglutinin, Surfactant pulmonary-associated D-binding protein
Host Species Rat
Immunogen E. Coli-derived recombinant mouse DMBT1, Trp215-Gly420, Accession # Q60997
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer
Primary or Secondary Primary
Gene ID (Entrez) 1755
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.