Learn More
Description
Specifications
Specifications
| Antigen | DMC1 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 1D12/4 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000, Immunoprecipitation reported in scientific literature |
| Formulation | Ascites with 0.1% Sodium Azide |
| Gene Accession No. | Q14565 |
| Gene Alias | disrupted meiotic cDNA1, yeast, homolog of, DMC1 dosage suppressor of mck1 homolog, meiosis-specific homologousrecombination (yeast), DMC1 homologue, HsLim15, LIM15yeast homolog) meiosis-specific homologousrecombination, meiotic recombination protein DMC1/LIM15 homolog, MGC150472, MGC150473 |
| Gene Symbols | DMC1 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
