Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DMC1 Antibody (1D12/4), Novus Biologicals™
SDP

Catalog No. NB100181SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100181SS 0.025 mL
NB100181 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB100181SS Supplier Novus Biologicals Supplier No. NB100181SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 1 publication

DMC1 Monoclonal specifically detects DMC1 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen DMC1
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 1D12/4
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunoprecipitation reported in scientific literature
Formulation Ascites with 0.1% Sodium Azide
Gene Accession No. Q14565
Gene Alias disrupted meiotic cDNA1, yeast, homolog of, DMC1 dosage suppressor of mck1 homolog, meiosis-specific homologousrecombination (yeast), DMC1 homologue, HsLim15, LIM15yeast homolog) meiosis-specific homologousrecombination, meiotic recombination protein DMC1/LIM15 homolog, MGC150472, MGC150473
Gene Symbols DMC1
Host Species Mouse
Immunogen This DMC1 Antibody (1D12/4) is made against His-tagged human DMC1, overexpressed in E. coli [UniProt# Q14565].
Molecular Weight of Antigen 37 kDa
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Cycle and Replication, Chromatin Research, DNA Repair, Homologous Recombination, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 11144
Test Specificity Human and mouse. Does not cross-react with human Rad51 in Western analysis.
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.